ENA448969307 Enamine

1-phenyl-4-(prop-2-yn-1-yl)piperazine

  • MDL number MFCD14631354
  • Cas Number : 21057-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214414
Availability details for
Quantity
  • Availability
  • Add to Cart