ENA448969308 Enamine

2-methoxy-N-[(4-methyl-1,3-thiazol-2-yl)methyl]aniline

  • MDL number MFCD19322068
  • Cas Number : 1267215-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214415
Availability details for
Quantity
  • Availability
  • Add to Cart