ENA448969321 Enamine

N-(3-aminophenyl)-2-(diethylamino)acetamide

  • MDL number MFCD09051849
  • Cas Number : 856047-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213351
Availability details for
Quantity
  • Availability
  • Add to Cart