ENA448969323 Enamine

2-(pyridin-4-ylmethoxy)acetic acid

  • MDL number MFCD20660938
  • Cas Number : 116882-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-213354
Availability details for
Quantity
  • Availability
  • Add to Cart