ENA448969329 Enamine

3,5-dichloro-4-[(dimethylcarbamoyl)methoxy]benzene-1-sulfonyl chloride

  • MDL number MFCD25830390
  • Cas Number : 1557293-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214513
Availability details for
Quantity
  • Availability
  • Add to Cart