ENA448969332 Enamine

2-(bromomethyl)-3-butyloxirane

  • MDL number MFCD20624749
  • Cas Number : 861320-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213356
Availability details for
Quantity
  • Availability
  • Add to Cart