ENA448969334 Enamine

propan-2-yl 5-(chlorosulfonyl)-2-methylbenzoate

  • MDL number MFCD21224845
  • Cas Number : 1486804-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214519
Availability details for
Quantity
  • Availability
  • Add to Cart