ENA448969337 Enamine

N-(pyridin-2-yl)pyridine-2-carbothioamide

  • MDL number MFCD00448857
  • Cas Number : 39122-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214523
Availability details for
Quantity
  • Availability
  • Add to Cart