ENA448969340 Enamine

(3-{[2-(dimethylamino)ethyl](methyl)amino}propyl)(methyl)amine

  • MDL number MFCD18867541
  • Cas Number : 1342437-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214529
Availability details for
Quantity
  • Availability
  • Add to Cart