ENA448969343 Enamine

2-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]acetic acid

  • MDL number MFCD16851376
  • Cas Number : 1250483-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213358
Availability details for
Quantity
  • Availability
  • Add to Cart