ENA448969346 Enamine

1-(difluoromethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD18089158
  • Cas Number : 899452-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214542
Availability details for
Quantity
  • Availability
  • Add to Cart