ENA448969347 Enamine

2-(5-aminopyridin-2-yl)ethan-1-ol

  • MDL number MFCD28505944
  • Cas Number : 1552277-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213359
Availability details for
Quantity
  • Availability
  • Add to Cart