ENA448969351 Enamine

1-(5-bromopyrazin-2-yl)pyrrolidin-2-one

  • MDL number MFCD10688564
  • Cas Number : 1027512-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214549
Availability details for
Quantity
  • Availability
  • Add to Cart