ENA448969365 Enamine

(5,6-dihydro-2H-pyran-3-yl)methanamine hydrochloride

  • MDL number MFCD28505946
  • Cas Number : 1803562-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213365
Availability details for
Quantity
  • Availability
  • Add to Cart