ENA448969372 Enamine

1-(bromomethyl)-1-fluorocyclohexane

  • MDL number MFCD20623965
  • Cas Number : 17171-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214593
Availability details for
Quantity
  • Availability
  • Add to Cart