ENA448969383 Enamine

4-(3,3-dimethylazetidin-2-yl)-1-methyl-1H-pyrazole dihydrochloride

  • MDL number MFCD28505950
  • Cas Number : 1803570-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213411
Availability details for
Quantity
  • Availability
  • Add to Cart