ENA448969400 Enamine

1-{thieno[3,2-c]pyridin-4-yl}piperazine

  • MDL number MFCD06796184
  • Cas Number : 106261-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214677
Availability details for
Quantity
  • Availability
  • Add to Cart