ENA448969406 Enamine

1-(2-methoxyphenyl)-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD16519188
  • Cas Number : 1282027-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213454
Availability details for
Quantity
  • Availability
  • Add to Cart