ENA448969408 Enamine

5-methyl-5-phenyl-1,3-oxazolidine-2,4-dione

  • MDL number MFCD20728046
  • Cas Number : 130689-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-76
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214687
Availability details for
Quantity
  • Availability
  • Add to Cart