ENA448969415 Enamine

2-(2-aminoethyl)-1H-1,3-benzodiazole-5-carboxylic acid dihydrochloride

  • MDL number MFCD09029283
  • Cas Number : 1187582-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214697
Availability details for
Quantity
  • Availability
  • Add to Cart