ENA448969426 Enamine

2-bromo-4-tert-butylbenzaldehyde

  • MDL number MFCD09029651
  • Cas Number : 246139-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214771
Availability details for
Quantity
  • Availability
  • Add to Cart