ENA448969428 Enamine

4-(propane-1-sulfonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD16093684
  • Cas Number : 1250742-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214794
Availability details for
Quantity
  • Availability
  • Add to Cart