ENA448969430 Enamine

3-ethoxy-2,2-dimethylpropan-1-amine

  • MDL number MFCD23733456
  • Cas Number : 1426955-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214797
Availability details for
Quantity
  • Availability
  • Add to Cart