ENA448969437 Enamine

6-fluoro-N-methylpyridin-2-amine

  • MDL number MFCD11520704
  • Cas Number : 325707-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214813
Availability details for
Quantity
  • Availability
  • Add to Cart