ENA448969443 Enamine

2,2-dimethyl-2,3,4,5-tetrahydro-1,4-benzoxazepine

  • MDL number MFCD26673252
  • Cas Number : 1519603-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214820
Availability details for
Quantity
  • Availability
  • Add to Cart