ENA448969445 Enamine

4-(propane-1-sulfonyl)benzene-1-sulfonamide

  • MDL number MFCD16093733
  • Cas Number : 98960-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214822
Availability details for
Quantity
  • Availability
  • Add to Cart