ENA448969452 Enamine

4-chloro-5-fluoro-2-(morpholin-4-yl)aniline

  • MDL number MFCD28506071
  • Cas Number : 1690822-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214840
Availability details for
Quantity
  • Availability
  • Add to Cart