ENA448969455 Enamine

2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD06411140
  • Cas Number : 194602-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214844
Availability details for
Quantity
  • Availability
  • Add to Cart