ENA448969456 Enamine

3-(4-bromophenyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12137232
  • Cas Number : 1153976-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214845
Availability details for
Quantity
  • Availability
  • Add to Cart