ENA448969458 Enamine

2,6-dichloro-4-iodopyridine-3-carboxylic acid

  • MDL number MFCD16658922
  • Cas Number : 343781-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-228
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214847
Availability details for
Quantity
  • Availability
  • Add to Cart