ENA448969459 Enamine

2-phenyl-1,3-dioxane-4-carboxylic acid

  • MDL number MFCD24483616
  • Cas Number : 178680-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214848
Availability details for
Quantity
  • Availability
  • Add to Cart