ENA448969465 Enamine

2-[5-(2,2,2-trifluoroethyl)-1,3,4-oxadiazol-2-yl]piperidine hydrochloride

  • MDL number MFCD28506076
  • Cas Number : 1803588-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214886
Availability details for
Quantity
  • Availability
  • Add to Cart