ENA448969468 Enamine

1-[(3-chlorophenyl)methyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD16328468
  • Cas Number : 1226226-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213500
Availability details for
Quantity
  • Availability
  • Add to Cart