ENA448969480 Enamine

ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-methylpropanoate

  • MDL number MFCD24623326
  • Cas Number : 76687-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213504
Availability details for
Quantity
  • Availability
  • Add to Cart