ENA448969504 Enamine

N-(4-acetyl-1,3-thiazol-2-yl)-3-aminobutanamide hydrochloride

  • MDL number MFCD28506086
  • Cas Number : 1803608-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214975
Availability details for
Quantity
  • Availability
  • Add to Cart