ENA448969506 Enamine

1-[(2-amino-6-fluorophenyl)methyl]-3-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD19474669
  • Cas Number : 1284791-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213517
Availability details for
Quantity
  • Availability
  • Add to Cart