ENA448969516 Enamine

naphthalene-2-carboximidamide

  • MDL number MFCD05663337
  • Cas Number : 5651-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215001
Availability details for
Quantity
  • Availability
  • Add to Cart