ENA448969517 Enamine

2-(2,2-dimethylpropyl)butanedioic acid

  • MDL number MFCD24513959
  • Cas Number : 1803601-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215002
Availability details for
Quantity
  • Availability
  • Add to Cart