ENA448969518 Enamine

2-(2-chlorophenyl)-1-methylpiperazine

  • MDL number MFCD10034704
  • Cas Number : 1018645-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213518
Availability details for
Quantity
  • Availability
  • Add to Cart