ENA448969520 Enamine

2-[4-(trifluoromethyl)pyridin-3-yl]acetic acid

  • MDL number MFCD11848109
  • Cas Number : 1227607-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215013
Availability details for
Quantity
  • Availability
  • Add to Cart