ENA448969521 Enamine

7-methyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD07377079
  • Cas Number : 1505-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215016
Availability details for
Quantity
  • Availability
  • Add to Cart