ENA448969525 Enamine

4-(2-amino-1-hydroxypropan-2-yl)phenol hydrochloride

  • MDL number MFCD28506093
  • Cas Number : 1803601-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215029
Availability details for
Quantity
  • Availability
  • Add to Cart