ENA448969531 Enamine

2,2-dimethyl-1-[2-(thiophen-3-yl)pyrrolidin-1-yl]propan-1-one

  • MDL number MFCD20089184
  • Cas Number : 1498742-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215039
Availability details for
Quantity
  • Availability
  • Add to Cart