ENA448969532 Enamine

2,2-dimethyl-1-[2-(pyridin-4-yl)pyrrolidin-1-yl]propan-1-one

  • MDL number MFCD20540801
  • Cas Number : 1378852-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215040
Availability details for
Quantity
  • Availability
  • Add to Cart