ENA448969533 Enamine

N-{[(2-chloro-6-fluorophenyl)methylidene]amino}guanidine

  • Cas Number : 1620238-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215041
Availability details for
Quantity
  • Availability
  • Add to Cart