ENA448969538 Enamine

3-tert-butyl-1H-1,2,4-triazole

  • MDL number MFCD11847493
  • Cas Number : 96440-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215054
Availability details for
Quantity
  • Availability
  • Add to Cart