ENA448969544 Enamine

[2-(4-ethylpiperazin-1-yl)ethyl](propyl)amine

  • MDL number MFCD14655202
  • Cas Number : 1211458-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215163
Availability details for
Quantity
  • Availability
  • Add to Cart