ENA448969545 Enamine

3-(dimethylamino)pyridine-2-carbonitrile

  • MDL number MFCD11656309
  • Cas Number : 97483-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215167
Availability details for
Quantity
  • Availability
  • Add to Cart