ENA448969546 Enamine

4-amino-3-(2,2-difluoroethoxy)benzamide

  • MDL number MFCD24280411
  • Cas Number : 1558390-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215168
Availability details for
Quantity
  • Availability
  • Add to Cart