ENA448969549 Enamine

[2-(2-methoxyethoxy)ethyl](methyl)amine

  • MDL number MFCD16107087
  • Cas Number : 124192-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215177
Availability details for
Quantity
  • Availability
  • Add to Cart